Learn More
Medchemexpress LLC Omadacycline (hydrochloride) | 1196800-39-1 | 99.5% | 593.11 | 1 G
SDP
Catalog No. 5000362099
Encompass_Preferred
Omadacycline hydrochloride (PTK 0796) is a first-in-class orally active aminomethylcycline antibacterial, belonging to the tetracycline class of antibiotics. It inhibits bacterial protein synthesis by binding to the 30S ribosomal subunit. This compound exhibits broad-spectrum antibacterial activity against aerobic and anaerobic Gram-positive and Gram-negative bacteria, as well as atypical bacteria. It is used in the research of acute bacterial skin and skin-structure infections, community-acquired pneumonia, and urinary tract infections.
  • Exhibits broad-spectrum antibacterial activity.
  • Effective against aerobic and anaerobic gram-positive and gram-negative bacteria.
  • Shows activity against atypical bacteria.
  • Inhibits bacterial protein synthesis by binding to the 30S ribosomal subunit.
  • Active against resistant strains including MRSA, VRE, beta-hemolytic streptococci, PRSP, and H. influenzae.
Catalog No. 50-003-62099 Supplier Medchemexpress LLC Supplier No. HY14865C1G
Omadacycline hydrochloride (PTK 0796) is a first-in-class orally active aminomethylcycline antibacterial, belonging to the tetracycline class of antibiotics. It inhibits bacterial protein synthesis by binding to the 30S ribosomal subunit. This compound exhibits broad-spectrum antibacterial activity against aerobic and anaerobic Gram-positive and Gram-negative bacteria, as well as atypical bacteria. It is used in the research of acute bacterial skin and skin-structure infections, community-acquired pneumonia, and urinary tract infections.
  • Exhibits broad-spectrum antibacterial activity.
  • Effective against aerobic and anaerobic gram-positive and gram-negative bacteria.
  • Shows activity against atypical bacteria.
  • Inhibits bacterial protein synthesis by binding to the 30S ribosomal subunit.
  • Active against resistant strains including MRSA, VRE, beta-hemolytic streptococci, PRSP, and H. influenzae.