Learn More
Medchemexpress LLC Omadacycline hydrochloride | 1196800-39-1 | >99.5% | 593.11 | 25 MG
SDP
Catalog No. 5000363921
Encompass_Preferred
Omadacycline hydrochloride is a first-in-class orally active aminomethylcycline antibacterial, belonging to the tetracycline class of antibiotics. It functions by inhibiting bacterial protein synthesis through binding to the 30S ribosomal subunit. It demonstrates broad-spectrum antibacterial activity against aerobic and anaerobic Gram-positive and Gram-negative bacteria, as well as atypical bacteria. This compound can be used for research into acute bacterial skin and skin-structure infections, community-acquired pneumonia, and urinary tract infections.
  • Inhibits bacterial protein synthesis by binding to the 30S ribosomal subunit.
  • Shows broad-spectrum antibacterial activity against various bacteria.
  • Used for research of acute bacterial skin and skin-structure infections.
  • Applicable for community-acquired pneumonia and urinary tract infections research.
Catalog No. 50-003-63921 Supplier Medchemexpress LLC Supplier No. 25MGHY14865C
Omadacycline hydrochloride is a first-in-class orally active aminomethylcycline antibacterial, belonging to the tetracycline class of antibiotics. It functions by inhibiting bacterial protein synthesis through binding to the 30S ribosomal subunit. It demonstrates broad-spectrum antibacterial activity against aerobic and anaerobic Gram-positive and Gram-negative bacteria, as well as atypical bacteria. This compound can be used for research into acute bacterial skin and skin-structure infections, community-acquired pneumonia, and urinary tract infections.
  • Inhibits bacterial protein synthesis by binding to the 30S ribosomal subunit.
  • Shows broad-spectrum antibacterial activity against various bacteria.
  • Used for research of acute bacterial skin and skin-structure infections.
  • Applicable for community-acquired pneumonia and urinary tract infections research.