Learn More
TARGETMOL CHEMICALS INC OMAVELOXOLONE 10MG
SDP
Catalog No. 502576414
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Omaveloxolone (RTA-408) (RTA-408) is a synthetic triterpenoid that activates the cytoprotective transcription factor Nrf2 and inhibits NF-(kappa)B signaling. Phase 2. purity: 98%
Catalog No. 50-257-6414 Supplier TARGETMOL CHEMICALS INC Supplier No. T691910MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Omaveloxolone (RTA-408) (RTA-408) is a synthetic triterpenoid that activates the cytoprotective transcription factor Nrf2 and inhibits NF-(kappa)B signaling. Phase 2. purity: 98%