Learn More
Medchemexpress LLC HY-12212 50mg Medchemexpress, Omaveloxolone CAS:1474034-05-3 Purity:>98%
SDP
Catalog No. 502028860
Encompass_Preferred
Medchemexpress, HY-12212 50mg Omaveloxolone CAS:1474034-05-3 Omaveloxolone (RTA 408) is an antioxidant inflammation modulator (AIM), which activates Nrf2 and suppresses nitric oxide (NO). Omaveloxolone attenuates osteoclastogenesis by inhibiting STING dependent NF-κb signaling. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8860 Supplier Medchemexpress LLC Supplier No. HY1221250MG
Medchemexpress, HY-12212 50mg Omaveloxolone CAS:1474034-05-3 Omaveloxolone (RTA 408) is an antioxidant inflammation modulator (AIM), which activates Nrf2 and suppresses nitric oxide (NO). Omaveloxolone attenuates osteoclastogenesis by inhibiting STING dependent NF-κb signaling. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.