Learn More
Selleck Chemical LLC Omaveloxolone5mg
SDP
Catalog No. 502173294
Encompass_Preferred
Omaveloxolone (RTA-408) is a synthetic triterpenoid that activates the cytoprotective transcription factor Nrf2 and inhibits NF-?B signaling. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3294 Supplier Selleck Chemical LLC Supplier No. S76725MG
Omaveloxolone (RTA-408) is a synthetic triterpenoid that activates the cytoprotective transcription factor Nrf2 and inhibits NF-?B signaling. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.