Learn More
Medchemexpress LLC HY-13997 10mg Medchemexpress, Ombitasvir CAS:1258226-87-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1399710MG
Encompass_Preferred
Medchemexpress, HY-13997 10mg Ombitasvir CAS:1258226-87-7 Ombitasvir is a potent inhibitor of the hepatitis C virus protein NS5A, with EC50s of 0.82 to 19.3 pM against HCV genotypes 1 to 5, and 366 pM against genotype 6a. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9045