Learn More
Selleck Chemical LLC Ombitasvir (ABT-267) S5403-5mg
SDP
Catalog No. 503109770
Encompass_Preferred
Ombitasvir (ABT-267) is an inhibitor of the HCV non-structural protein 5A with antiviral activity
Catalog No. 50-310-9770 Supplier Selleck Chemical LLC Supplier No. S54035MG
Ombitasvir (ABT-267) is an inhibitor of the HCV non-structural protein 5A with antiviral activity