Learn More
Medchemexpress LLC HY-13997 100mg , Ombitasvir ABT-267 CAS:1258226-87-7 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY13997100MG
Encompass_Preferred
Medchemexpress, HY-13997 100mg Ombitasvir ABT-267 CAS:1258226-87-7 Formula:C50H67N7O8 EC50: 0.82 to 19.3 pM (HCV genotypes 1 to 5), 366 pM (HCV genotype 6a) Purity:98% Ombitasvir is a potent inhibitor of the hepatitis C virus protein NS5A , with EC 50 s of 0.82 to 19.3 pM against HCV genotypes 1 to 5 and 366 pM against genotype 6a. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3105