Learn More
Medchemexpress LLC HY-18256 5mg Medchemexpress, Ombrabulin (hydrochloride) CAS:253426-24-3 Purity:>98%
SDP
Catalog No. 502082029
Encompass_Preferred
Medchemexpress, HY-18256 5mg Ombrabulin (hydrochloride) CAS:253426-24-3 Ombrabulin hydrochloride is a derivative of CA-4 phosphate, which is known to exhibit antivascular effects through selective disruption of the tubulin cytoskeleton of endothelial cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2029 Supplier Medchemexpress LLC Supplier No. HY182565MG
Medchemexpress, HY-18256 5mg Ombrabulin (hydrochloride) CAS:253426-24-3 Ombrabulin hydrochloride is a derivative of CA-4 phosphate, which is known to exhibit antivascular effects through selective disruption of the tubulin cytoskeleton of endothelial cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.