Learn More
Medchemexpress LLC Ombrabulin hydrochloride | 253426-24-3 | 99.4% | 438.90 | 100 MG
SDP
Catalog No. 5000387621
Encompass_Preferred
Ombrabulin hydrochloride is a derivative of CA-4 phosphate, known to exhibit antivascular effects through selective disruption of the tubulin cytoskeleton of endothelial cells. It acts as a tubulin inhibitor, demonstrating potent cytotoxicity against various cancer cell lines and showing efficacy in reducing tumor weight in vivo.
  • Acts as a tubulin inhibitor
  • Exhibits antivascular effects
  • Disrupts the tubulin cytoskeleton of endothelial cells
  • Shows cytotoxicity against DU-145, HeLa, MCF7, NCI-H460, and SK-OV-3 cell lines
  • Proven effective in reducing tumor weight in animal models
  • Tested in clinical trials for solid tumors and various cancers
Catalog No. 50-003-87621 Supplier Medchemexpress LLC Supplier No. HY18256100MG
Ombrabulin hydrochloride is a derivative of CA-4 phosphate, known to exhibit antivascular effects through selective disruption of the tubulin cytoskeleton of endothelial cells. It acts as a tubulin inhibitor, demonstrating potent cytotoxicity against various cancer cell lines and showing efficacy in reducing tumor weight in vivo.
  • Acts as a tubulin inhibitor
  • Exhibits antivascular effects
  • Disrupts the tubulin cytoskeleton of endothelial cells
  • Shows cytotoxicity against DU-145, HeLa, MCF7, NCI-H460, and SK-OV-3 cell lines
  • Proven effective in reducing tumor weight in animal models
  • Tested in clinical trials for solid tumors and various cancers