Learn More
Medchemexpress LLC Ombrabulin hydrochloride | 253426-24-3 | 99.43 % | 438.90 | 10 MM 1 ML
SDP
Supplier:  Medchemexpress LLC HY1825610MM/1ML
Encompass_Preferred
Ombrabulin hydrochloride is a derivative of CA-4 phosphate, known for its antivascular effects achieved by selectively disrupting the tubulin cytoskeleton of endothelial cells.
  • Exhibits antivascular effects.
  • Selectively disrupts the tubulin cytoskeleton of endothelial cells.
  • Is a derivative of CA-4 phosphate.
Catalog No. 50-003-87620