Learn More
Cayman Chemical OmbrabulInhydrochlorIde 10mg
SDP
Catalog No. 502912754
Encompass_Preferred
A tubulin polymerization inhibitor (IC50 = 11.3 µM) and tumor vascular disrupting agent; induces cell cycle arrest at the G2/M phase and apoptosis in HEY A8 and mouse mesenteric endothelial cells; reduces tumor growth and induces tumor necrosis in a HEY A8 mouse xenograft model at 30 mg/kg
Catalog No. 50-291-2754 Supplier Cayman Chemical Supplier No. 3489710 MG
A tubulin polymerization inhibitor (IC50 = 11.3 µM) and tumor vascular disrupting agent; induces cell cycle arrest at the G2/M phase and apoptosis in HEY A8 and mouse mesenteric endothelial cells; reduces tumor growth and induces tumor necrosis in a HEY A8 mouse xenograft model at 30 mg/kg