Learn More
Medchemexpress LLC Ombuin | 529-40-8 | 1 MG
SDP
Catalog No. 5000420959
Encompass_Preferred
Ombuin is a compound isolated from Zanthoxylum armatum, known for its broad-spectrum antibacterial effects with MIC ranges from 125 to 500 μg/mL. This product is intended for research use only and should be handled by qualified personnel in appropriately equipped facilities.
  • Isolated from Zanthoxylum armatum
  • Exhibits broad-spectrum antibacterial effects
  • Effective against bacteria with MIC ranges from 125 to 500 μg/mL
  • Classified as a flavonol, flavonoid, phenol, polyphenol
  • Appears as a solid
  • Melting point of 229-230°C
Catalog No. 50-004-20959 Supplier Medchemexpress LLC Supplier No. HYN31391MG
Ombuin is a compound isolated from Zanthoxylum armatum, known for its broad-spectrum antibacterial effects with MIC ranges from 125 to 500 μg/mL. This product is intended for research use only and should be handled by qualified personnel in appropriately equipped facilities.
  • Isolated from Zanthoxylum armatum
  • Exhibits broad-spectrum antibacterial effects
  • Effective against bacteria with MIC ranges from 125 to 500 μg/mL
  • Classified as a flavonol, flavonoid, phenol, polyphenol
  • Appears as a solid
  • Melting point of 229-230°C