Learn More
Medchemexpress LLC Ombuin | 529-40-8 | 5 MG
SDP
Catalog No. 5000421610
Encompass_Preferred
Ombuin is a compound isolated from *Zanthoxylum armatum*, exhibiting broad-spectrum antibacterial effects with MIC ranges from 125 to 500 μg/mL. It also demonstrates anticancer activity against various human cell lines and inhibits LPS-induced nitric oxide production. This compound is categorized under flavonoids, flavonols, phenols, and polyphenols.
  • Isolated from *Zanthoxylum armatum*
  • Displays broad-spectrum antibacterial effects
  • Exhibits anticancer activity against human cell lines
  • Inhibits LPS-induced nitric oxide production
  • Categorized as a flavonoid, flavonol, phenol, and polyphenol
  • Solid appearance with white to yellow color
  • Stable under recommended storage conditions
  • For research use only
Catalog No. 50-004-21610 Supplier Medchemexpress LLC Supplier No. HYN31395MG
Ombuin is a compound isolated from *Zanthoxylum armatum*, exhibiting broad-spectrum antibacterial effects with MIC ranges from 125 to 500 μg/mL. It also demonstrates anticancer activity against various human cell lines and inhibits LPS-induced nitric oxide production. This compound is categorized under flavonoids, flavonols, phenols, and polyphenols.
  • Isolated from *Zanthoxylum armatum*
  • Displays broad-spectrum antibacterial effects
  • Exhibits anticancer activity against human cell lines
  • Inhibits LPS-induced nitric oxide production
  • Categorized as a flavonoid, flavonol, phenol, and polyphenol
  • Solid appearance with white to yellow color
  • Stable under recommended storage conditions
  • For research use only