Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ombuoside | 20188-85-6 | 5 MG
SDP

Catalog No. 5000421586
Encompass_Preferred

Ombuoside is a compound with antioxidant properties, capable of inhibiting ROS production and apoptosis. It exhibits neuroprotective effects by acting through the ERK-JNK-caspase-3 system and promotes Dopamine biosynthesis via TH and CREB activation. Additionally, Ombuoside shows antimicrobial activity against various Gram-positive and Gram-negative bacteria, as well as Candida albicans.

  • Antioxidant: Inhibits ROS production and apoptosis.
  • Neuroprotective: Acts via the ERK-JNK-caspase-3 system.
  • Dopamine biosynthesis: Promotes Dopamine biosynthesis through TH and CREB activation.
  • Antimicrobial: Exhibits activity against several Gram-positive and Gram-negative bacteria, and Candida albicans.
  • Purity: 99.68%.
  • Appearance: White to light yellow solid.
  • Research use only.

Catalog No. 50-004-21586 Supplier Medchemexpress LLC Supplier No. HYN31385MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ombuoside is a compound with antioxidant properties, capable of inhibiting ROS production and apoptosis. It exhibits neuroprotective effects by acting through the ERK-JNK-caspase-3 system and promotes Dopamine biosynthesis via TH and CREB activation. Additionally, Ombuoside shows antimicrobial activity against various Gram-positive and Gram-negative bacteria, as well as Candida albicans.

  • Antioxidant: Inhibits ROS production and apoptosis.
  • Neuroprotective: Acts via the ERK-JNK-caspase-3 system.
  • Dopamine biosynthesis: Promotes Dopamine biosynthesis through TH and CREB activation.
  • Antimicrobial: Exhibits activity against several Gram-positive and Gram-negative bacteria, and Candida albicans.
  • Purity: 99.68%.
  • Appearance: White to light yellow solid.
  • Research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.