Learn More
TARGETMOL CHEMICALS INC OMDM-6 5MG
SDP
Catalog No. 502581469
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. OMDM-6 is a dual agonist of TRPV1(EC50 = 75 nM) and CB1 (Ki = 3.2 uM). OMDM-6 inhibits anandamide cellular uptake with a Ki of 7.0 uM. purity: 99%
Catalog No. 50-258-1469 Supplier TARGETMOL CHEMICALS INC Supplier No. T123075MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. OMDM-6 is a dual agonist of TRPV1(EC50 = 75 nM) and CB1 (Ki = 3.2 uM). OMDM-6 inhibits anandamide cellular uptake with a Ki of 7.0 uM. purity: 99%