Learn More
Cayman Chemical OmecamtIv MecarbIl 10mg
SDP
Catalog No. 502982361
Encompass_Preferred
A cardiac myosin ATPase activator (AC40 = 0.58 µM for the bovine enzyme); increases fractional shortening in rats in a dose-dependent manner
Catalog No. 50-298-2361 Supplier Cayman Chemical Supplier No. 2107410 MG
A cardiac myosin ATPase activator (AC40 = 0.58 µM for the bovine enzyme); increases fractional shortening in rats in a dose-dependent manner