Learn More
Selleck Chemical LLC Omecamtiv mecarbil (CK-1827452) 50mg 873697-71-3
SDP
Supplier:  Selleck Chemical LLC S262350MG
Encompass_Preferred
Omecamtiv mecarbil (CK-1827452) is a specific cardiac myosin activator and a clinical drug for left ventricular systolic heart failure. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-546-6