Learn More
Medchemexpress LLC Omega-3 arachidonic acid ethyl ester | 123940-93-2 | 98.0% | 332.52 | C22H36O2 | 1MG
SDP
Catalog No. 5000238752
Encompass_Preferred
Omega-3 arachidonic acid ethyl ester is the ethyl-ester derivative of an omega-3 polyunsaturated fatty acid supplied in ethanol at high purity. It provides increased lipophilicity compared with the free acid and is intended for lipid research, formulation, and analytical applications. Molecular formula C22H36O2; molecular weight 332.52 g/mol. Available in small vial sizes suitable for analytical and preparative use.
  • Supplied in ethanol at 98.0% purity.
  • Ethyl ester increases lipophilicity for formulation and metabolism studies.
  • Suitable for analytical and preparative lipid research.
  • Molecular formula C22H36O2; molecular weight 332.52 g/mol.
  • Available in small vial sizes for laboratory use.
Catalog No. 50-002-38752 Supplier Medchemexpress LLC Supplier No. HY1251391MG
Omega-3 arachidonic acid ethyl ester is the ethyl-ester derivative of an omega-3 polyunsaturated fatty acid supplied in ethanol at high purity. It provides increased lipophilicity compared with the free acid and is intended for lipid research, formulation, and analytical applications. Molecular formula C22H36O2; molecular weight 332.52 g/mol. Available in small vial sizes suitable for analytical and preparative use.
  • Supplied in ethanol at 98.0% purity.
  • Ethyl ester increases lipophilicity for formulation and metabolism studies.
  • Suitable for analytical and preparative lipid research.
  • Molecular formula C22H36O2; molecular weight 332.52 g/mol.
  • Available in small vial sizes for laboratory use.