Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC Atrix Industrial Vacuum, Cordless, 22V, ULPA Filtration, 1 gal

Catalog No. 502837987
Encompass

This industrial vacuum is designed for cleaning hazardous debris applications such as asbestos, lead, silica dust, and mold, or in other environments requiring an IEC/EN 60335-2-69, Annex AA-Class H certified vacuum cleaner. It features cordless operation for portability and ULPA filtration for effective containment of harmful particles.

  • Cordless vacuum operation for enhanced mobility.
  • Patented SafeGuard 360 H14 ULPA filtration.
  • Full filter indicator light.
  • Permanently attached heavy duty filter retention plug.
  • Self-storage and contained accessories.
  • Powerful 22V DC brushless digital motor.
  • 30+ minute run time.
  • One-gallon SafeGuard 360 H14 ULPA filter with 99.999% efficiency at .12 micron.
  • Toolbox structure for convenient use, transport, and storage.

Catalog No. 50-283-7987 Supplier MSC Supplier No. 82990086
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This industrial vacuum is designed for cleaning hazardous debris applications such as asbestos, lead, silica dust, and mold, or in other environments requiring an IEC/EN 60335-2-69, Annex AA-Class H certified vacuum cleaner. It features cordless operation for portability and ULPA filtration for effective containment of harmful particles.

  • Cordless vacuum operation for enhanced mobility.
  • Patented SafeGuard 360 H14 ULPA filtration.
  • Full filter indicator light.
  • Permanently attached heavy duty filter retention plug.
  • Self-storage and contained accessories.
  • Powerful 22V DC brushless digital motor.
  • 30+ minute run time.
  • One-gallon SafeGuard 360 H14 ULPA filter with 99.999% efficiency at .12 micron.
  • Toolbox structure for convenient use, transport, and storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.