Learn More
Omega Biotek Inc Plasmid DNA Mini Kit I, (V-spin), 50 preps
Catalog No. NC0310358
Encompass
Isolate up to 25 ug plasmid DNA from 1-5 mL bacterial cultures using mini spin columns.
Catalog No. NC0310358 Supplier Omega Biotek Inc Supplier No. D694301
Isolate up to 25 ug plasmid DNA from 1-5 mL bacterial cultures using mini spin columns.