Learn More
Cytiva (Formerly Pall Lab) Nanosep Centrifugal Devices with Omega™ Membrane - 300K, orange (24/pkg)
Supplier:  Cytiva OD300C33
Encompass
Bio-Inert modified nylon membranes exhibit 25-50% lower binding than traditional nylon membranes This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-197-9587