Learn More
MSC Atrix Backpack Vacuum, HEPA Filtration, 1.00 gal Capacity, 69 dB
Catalog No. 502836933
Encompass
The Omega SafeTech HEPA Backpack Vacuum is engineered for dry hazardous particulate cleaning. It operates quietly at 69 decibels and features a SafeGuard 360 True HEPA Filtration Cartridge, which is rated 99.97% efficient at 0.3 microns. Its design simplifies filter replacements with one quick cartridge changeout, making it suitable for demanding environments.
  • Engineered for dry hazardous particulate cleaning
  • Operates quietly at 69 decibels
  • Features true HEPA filtration, rated 99.97% efficient at 0.3 microns
  • User-friendly design for quick filter replacements
  • Equipped with a comprehensive set of accessories
  • Ideal for asbestos, lead dust, silica dust, mold, and bed bugs
Catalog No. 50-283-6933 Supplier MSC Supplier No. 60236635
The Omega SafeTech HEPA Backpack Vacuum is engineered for dry hazardous particulate cleaning. It operates quietly at 69 decibels and features a SafeGuard 360 True HEPA Filtration Cartridge, which is rated 99.97% efficient at 0.3 microns. Its design simplifies filter replacements with one quick cartridge changeout, making it suitable for demanding environments.
  • Engineered for dry hazardous particulate cleaning
  • Operates quietly at 69 decibels
  • Features true HEPA filtration, rated 99.97% efficient at 0.3 microns
  • User-friendly design for quick filter replacements
  • Equipped with a comprehensive set of accessories
  • Ideal for asbestos, lead dust, silica dust, mold, and bed bugs