Learn More
MSC Shepherd Caster Omega Series Swivel Caster, 3 in. Rubber Wheel, Pedal Brake, 1 ea
Catalog No. 5000513041
Encompass
The Omega Series swivel caster is designed for reliable mobility in various professional applications. It features a 3 in. diameter black non-marking rubber wheel with a 0.9375 in. width, engineered to protect surfaces while ensuring smooth operation. This unit includes an integrated pedal brake to secure equipment in place when needed.
  • Swivel design with integrated pedal brake for versatility
  • Precision ball bearing construction for smooth movement
  • Black non-marking rubber wheel prevents surface damage
  • Includes protective swivel seals
  • Threaded stem mounting type with 0.5 in. x 1.5 in. stem size
  • Dynamic capacity rating of 110 lbs.
Catalog No. 50-005-13041 Supplier MSC Supplier No. 74582859
The Omega Series swivel caster is designed for reliable mobility in various professional applications. It features a 3 in. diameter black non-marking rubber wheel with a 0.9375 in. width, engineered to protect surfaces while ensuring smooth operation. This unit includes an integrated pedal brake to secure equipment in place when needed.
  • Swivel design with integrated pedal brake for versatility
  • Precision ball bearing construction for smooth movement
  • Black non-marking rubber wheel prevents surface damage
  • Includes protective swivel seals
  • Threaded stem mounting type with 0.5 in. x 1.5 in. stem size
  • Dynamic capacity rating of 110 lbs.