Learn More
Research Products International Corp Omega Zen Pipettor, Gray, Quickly Fills 25, 50 and 100 ml Pipettes
SDP
Catalog No. 501977170
Encompass
Omega Pipettor quickly fills 25, 50 and 100 ml pipettes. Offers powerful performance and superior fingertip control. Fits pipettes from 1 - 100 ml and dispenses slowly and gently to protect valuable samples. Features UV resistant housing. Unit is rechargeable during use and features quiet motor and five bar LED battery light. This item is supplied with a sterile filter, battery charger, wall mount and instructions.
Catalog No. 50-197-7170 Supplier Research Products International Corp Supplier No. 353000GR
Add to Cart
Add to Cart
Omega Pipettor quickly fills 25, 50 and 100 ml pipettes. Offers powerful performance and superior fingertip control. Fits pipettes from 1 - 100 ml and dispenses slowly and gently to protect valuable samples. Features UV resistant housing. Unit is rechargeable during use and features quiet motor and five bar LED battery light. This item is supplied with a sterile filter, battery charger, wall mount and instructions.