Learn More
Zen Bio Inc Omental Adipocytes 96-Well Plate, BMI >30.0
SDP
Catalog No. NC1784960
Encompass
The greater omentum is the largest peritoneal fold within the abdomen and has an immunologic function. The omentum aids in isolating peritoneal infection and absorbing contaminants through the mesothelial stomata. Adipokines and fatty acids from this depot have direct access to the liver through the portal circulation which may lead to hepatic dysfunction.
Catalog No. NC1784960 Supplier Zen Bio Inc Supplier No. OA10963
The greater omentum is the largest peritoneal fold within the abdomen and has an immunologic function. The omentum aids in isolating peritoneal infection and absorbing contaminants through the mesothelial stomata. Adipokines and fatty acids from this depot have direct access to the liver through the portal circulation which may lead to hepatic dysfunction.