Learn More
Enzo Life Sciences Omentin 1 (human), (recombinant) (10µg)
SDP
Supplier:  Enzo Life Sciences ALX522108C010
Encompass_Preferred
Has no effect on basal glucose uptake but enhances insulin-stimulated glucose uptake in adipocytes. Increases AKT phosphorylation in the absence and presence of insulin. May play a role in the defense system against microorganisms. May specifically recognize carbohydrate chains of pathogens and bacterial components containing galactofuranosyl residues, in a calcium-dependent manner. May be involved in iron metabolism. Has no effect on basal glucose uptake but enhances insulin-stimulated glucose uptake in adipocytes. Increases AKT phosphorylation in the absence and presence of insulin. Alternative name: Intelectin 1, Endothelial lectin HL-1, Intestinal lactoferrin receptor, Galactofuranose binding lectin. Concentration: 0.1mg/ml after reconstitution. Purity: ≥95% (SDS-PAGE). Formulation: Lyophilized. Contains PBS. Source: Produced in HEK 293 cells. Human omentin 1 (intelectin-1) (aa 17-313) is fused at the N-terminus to a FLAG-tag.
Catalog No. 50-200-9428