Learn More
AdipoGen Omentin (human) (rec.) (10 µg)
SDP
Supplier:  AdipoGen AG40B0042C010
Encompass_Preferred
Protein. Human omentin (aa 17-313) is fused at the N-terminus to a FLAG-tag. Source: CHO cells. Endotoxin content: <0.1EU/µg purified protein (LAL test; Lonza). Lyophilized. Contains PBS. Purity: >95% (SDS-PAGE). Omentin (Intelectin-1) is highly expressed in omental adipose tissue, small intestine and various other tissues. Omentin has no effect on basal glucose uptake but enhances insulin-stimulated glucose uptake in adipocytes. It increases AKT phosphorylation in the absence and presence of insulin. It was suggested to play a role in the defense system against microorganisms by recognizing carbohydrate chains of pathogens and bacterial components containing galactofuranosyl residues.
Catalog No. 50-205-3118