Learn More
AdipoGen Omentin human pAb
SDP
Supplier:  AdipoGen AG25A0051C100
Encompass_Preferred
Polyclonal Antibody. Recognizes human omentin. Detects a band of ~40kDa by Western blot. Source Rabbit. Applications ELISA, WB. Liquid. 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives. Omentin Intelectin-1 is highly expressed in omental adipose tissue, small intestine and various other tissues. Omentin has no effect on basal glucose uptake but enhances insulin-stimulated glucose uptake in adipocytes. It increases AKT phosphorylation in the absence and presence of insulin. It was suggested to play a role in the defense system against microorganisms by recognizing carbohydrate chains of pathogens and bacterial components containing galactofuranosyl residues.
Catalog No. 50-256-3261