Learn More
Enzo Life Sciences Omentin (rat), (recombinant) (10µg)
SDP
Supplier:  Enzo Life Sciences ALX522132C010
Encompass_Preferred
Alternative name: Intelectin 1, Endothelial lectin HL-1, Intestinal lactoferrin receptor, Galactofuranose binding lectin. Concentration: 0.1mg/ml after reconstitution. Purity: ≥95% (SDS-PAGE). Formulation: Lyophilized. Contains PBS. Source: Produced in HEK 293 cells. Rat omentin (aa 20-313) is fused at the N-terminus to a FLAG-tag. Long Term Storage: -20°C.
Catalog No. 50-200-9446