Learn More
Apexbio Technology LLC Omeprazole 73590-58-6 100mg
SDP
Catalog No. 502240107
Encompass_Preferred
Omeprazole (73590-58-6) is a small-molecule inhibitor targeting H K -ATPase an enzyme essential for gastric acid secretion It is designed to inhibit the proton pump thereby reducing active transport of H ions into the gastric lumen Omeprazole exerts its biological activity primarily through irreversible binding to the H K -ATPase enzyme In in vitro studies Omeprazole demonstrates inhibitory activity with an IC value of approximately 5 8 M against purified H K -ATPase and an IC of 0 16 M in assays evaluating histamine-induced gastric acid production Based on these pharmacological properties Omeprazole holds research potential in the investigation of acid-mediated gastrointestinal conditions and related pathways
This item is not returnable. View return policy
Catalog No. 50-224-0107 Supplier Apexbio Technology LLC Supplier No. A2845100
Omeprazole (73590-58-6) is a small-molecule inhibitor targeting H K -ATPase an enzyme essential for gastric acid secretion It is designed to inhibit the proton pump thereby reducing active transport of H ions into the gastric lumen Omeprazole exerts its biological activity primarily through irreversible binding to the H K -ATPase enzyme In in vitro studies Omeprazole demonstrates inhibitory activity with an IC value of approximately 5 8 M against purified H K -ATPase and an IC of 0 16 M in assays evaluating histamine-induced gastric acid production Based on these pharmacological properties Omeprazole holds research potential in the investigation of acid-mediated gastrointestinal conditions and related pathways