Learn More
Medchemexpress LLC Omeprazole-13C,d3 | 1261395-28-1 | 98.0% | C1613CH16D3N3O3S | 1 MG
SDP
Catalog No. 5000432892
Encompass_Preferred
Omeprazole-13C,d3 is a 13C-labeled and deuterium labeled Omeprazole, which is a proton pump inhibitor (PPI) used for acid-related gastrointestinal disorders. It competitively inhibits CYP2C19 activity and also inhibits the growth of Gram-positive and Gram-negative bacteria. It is also a potent brain penetrant neutral sphingomyelinase (N-SMase) inhibitor (exosome inhibitor).
  • 13C-labeled and deuterium labeled Omeprazole.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-32892 Supplier Medchemexpress LLC Supplier No. HYB0113S31MG
Omeprazole-13C,d3 is a 13C-labeled and deuterium labeled Omeprazole, which is a proton pump inhibitor (PPI) used for acid-related gastrointestinal disorders. It competitively inhibits CYP2C19 activity and also inhibits the growth of Gram-positive and Gram-negative bacteria. It is also a potent brain penetrant neutral sphingomyelinase (N-SMase) inhibitor (exosome inhibitor).
  • 13C-labeled and deuterium labeled Omeprazole.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.