Learn More
Medchemexpress LLC Omeprazole-13C,d3 | 1261395-28-1 | 98.0% | C1613CH16D3N3O3S | 5 MG
SDP
Catalog No. 5000432130
Encompass_Preferred
Omeprazole-13C,d3 is a 13C-labeled and deuterium labeled proton pump inhibitor (PPI). It is used for treating acid-related gastrointestinal disorders and competitively inhibits CYP2C19 activity. This compound also inhibits the growth of Gram-positive and Gram-negative bacteria and acts as a potent brain penetrant neutral sphingomyelinase (N-SMase) inhibitor.
  • 13C-labeled and deuterium labeled
  • Proton pump inhibitor (PPI)
  • Treats acid-related gastrointestinal disorders
  • Competitively inhibits CYP2C19 activity
  • Inhibits Gram-positive and Gram-negative bacteria
  • Potent brain penetrant neutral sphingomyelinase (N-SMase) inhibitor
  • Suitable as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-32130 Supplier Medchemexpress LLC Supplier No. HYB0113S35MG
Add to Cart
Add to Cart
Omeprazole-13C,d3 is a 13C-labeled and deuterium labeled proton pump inhibitor (PPI). It is used for treating acid-related gastrointestinal disorders and competitively inhibits CYP2C19 activity. This compound also inhibits the growth of Gram-positive and Gram-negative bacteria and acts as a potent brain penetrant neutral sphingomyelinase (N-SMase) inhibitor.
  • 13C-labeled and deuterium labeled
  • Proton pump inhibitor (PPI)
  • Treats acid-related gastrointestinal disorders
  • Competitively inhibits CYP2C19 activity
  • Inhibits Gram-positive and Gram-negative bacteria
  • Potent brain penetrant neutral sphingomyelinase (N-SMase) inhibitor
  • Suitable as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS