Learn More
Selleck Chemical LLC Omeprazole 50mg 73590-58-6
SDP
Supplier:  Selleck Chemical LLC S138950MG
Encompass_Preferred
Omeprazole is a proton pump inhibitor that blocks H(+)-K(+)-ATPase, used to treat dyspepsia, peptic ulcer disease, gastroesophageal reflux disease, laryngopharyngeal reflux, and Zollinger¿Ellison syndrome. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1725437