Learn More
Medchemexpress LLC Omeprazole | 73590-58-6 | 100.0% | C17H19N3O3S | 1 ML
SDP
Catalog No. 5000432291
Encompass_Preferred
Omeprazole is a proton pump inhibitor (PPI) used for treating acid-related gastrointestinal disorders. It competitively inhibits CYP2C19 activity and inhibits the growth of Gram-positive and Gram-negative bacteria. Additionally, it is a potent brain-penetrant neutral sphingomyelinase (N-SMase) inhibitor, functioning as an exosome inhibitor.
  • Proton pump inhibitor (PPI)
  • Used for treating acid-related gastrointestinal disorders
  • Competitively inhibits CYP2C19 activity
  • Inhibits the growth of Gram-positive and Gram-negative bacteria
  • Potent brain-penetrant neutral sphingomyelinase (N-SMase) inhibitor
  • Functions as an exosome inhibitor
  • Intended for research use only
Catalog No. 50-004-32291 Supplier Medchemexpress LLC Supplier No. HYB011310MM/1ML
Omeprazole is a proton pump inhibitor (PPI) used for treating acid-related gastrointestinal disorders. It competitively inhibits CYP2C19 activity and inhibits the growth of Gram-positive and Gram-negative bacteria. Additionally, it is a potent brain-penetrant neutral sphingomyelinase (N-SMase) inhibitor, functioning as an exosome inhibitor.
  • Proton pump inhibitor (PPI)
  • Used for treating acid-related gastrointestinal disorders
  • Competitively inhibits CYP2C19 activity
  • Inhibits the growth of Gram-positive and Gram-negative bacteria
  • Potent brain-penetrant neutral sphingomyelinase (N-SMase) inhibitor
  • Functions as an exosome inhibitor
  • Intended for research use only