Learn More
Medchemexpress LLC Omeprazole | 73590-58-6 | 100.0% | C17H19N3O3S | 10 G
SDP
Catalog No. 5000432794
Encompass_Preferred
Omeprazole is a proton pump inhibitor (PPI) primarily used for treating acid-related gastrointestinal disorders. It competitively inhibits CYP2C19 activity, inhibits the growth of both Gram-positive and Gram-negative bacteria, and acts as a potent brain penetrant neutral sphingomyelinase (N-SMase) inhibitor.
  • Treats dyspepsia, peptic ulcer disease, and gastroesophageal reflux disease.
  • Effective for laryngopharyngeal reflux and Zollinger-Ellison syndrome.
  • Inhibits gastric acid secretion.
  • Competitively inhibits CYP2C19 activity.
  • Inhibits growth of Gram-positive and Gram-negative bacteria.
  • Functions as a brain penetrant neutral sphingomyelinase (N-SMase) inhibitor.
Catalog No. 50-004-32794 Supplier Medchemexpress LLC Supplier No. HYB011310G
Omeprazole is a proton pump inhibitor (PPI) primarily used for treating acid-related gastrointestinal disorders. It competitively inhibits CYP2C19 activity, inhibits the growth of both Gram-positive and Gram-negative bacteria, and acts as a potent brain penetrant neutral sphingomyelinase (N-SMase) inhibitor.
  • Treats dyspepsia, peptic ulcer disease, and gastroesophageal reflux disease.
  • Effective for laryngopharyngeal reflux and Zollinger-Ellison syndrome.
  • Inhibits gastric acid secretion.
  • Competitively inhibits CYP2C19 activity.
  • Inhibits growth of Gram-positive and Gram-negative bacteria.
  • Functions as a brain penetrant neutral sphingomyelinase (N-SMase) inhibitor.