Learn More
Medchemexpress LLC Omeprazole | 73590-58-6 | 100.0% | C17H19N3O3S | 25 G
SDP
Catalog No. 5000358952
Encompass_Preferred
Omeprazole is a proton pump inhibitor (PPI) used for treating acid-related gastrointestinal disorders, including dyspepsia, peptic ulcer disease, gastroesophageal reflux disease, laryngopharyngeal reflux, and Zollinger-Ellison syndrome.
  • Used for treating acid-related gastrointestinal disorders
  • Competitively inhibits CYP2C19 activity with a Ki of 2 to 6 μM
  • Inhibits the growth of both Gram-positive and Gram-negative bacteria
  • Potent brain-penetrant neutral sphingomyelinase (N-SMase) inhibitor (exosome inhibitor)
Catalog No. 50-003-58952 Supplier Medchemexpress LLC Supplier No. 25GHYB0113
Omeprazole is a proton pump inhibitor (PPI) used for treating acid-related gastrointestinal disorders, including dyspepsia, peptic ulcer disease, gastroesophageal reflux disease, laryngopharyngeal reflux, and Zollinger-Ellison syndrome.
  • Used for treating acid-related gastrointestinal disorders
  • Competitively inhibits CYP2C19 activity with a Ki of 2 to 6 μM
  • Inhibits the growth of both Gram-positive and Gram-negative bacteria
  • Potent brain-penetrant neutral sphingomyelinase (N-SMase) inhibitor (exosome inhibitor)