Learn More
Medchemexpress LLC Omeprazole | 73590-58-6 | 99.86% | C17H19N3O3S | 100 MG
SDP
Catalog No. 5000433517
Encompass_Preferred
Omeprazole (Standard) is an analytical standard intended for research and analytical applications. It serves as a reference standard in qualitative, quantitative, and methodological research experiments, including HPLC, GC, and MS. This compound acts as a proton pump inhibitor (PPI) and can inhibit the growth of Gram-positive and Gram-negative bacteria.
  • Reference standard for analytical applications
  • Suitable for HPLC, GC, and MS experiments
  • Functions as a proton pump inhibitor (PPI)
  • Inhibits growth of Gram-positive and Gram-negative bacteria
  • Potent brain penetrant neutral sphingomyelinase (N-SMase) inhibitor
Catalog No. 50-004-33517 Supplier Medchemexpress LLC Supplier No. HYB0113R100MG
Omeprazole (Standard) is an analytical standard intended for research and analytical applications. It serves as a reference standard in qualitative, quantitative, and methodological research experiments, including HPLC, GC, and MS. This compound acts as a proton pump inhibitor (PPI) and can inhibit the growth of Gram-positive and Gram-negative bacteria.
  • Reference standard for analytical applications
  • Suitable for HPLC, GC, and MS experiments
  • Functions as a proton pump inhibitor (PPI)
  • Inhibits growth of Gram-positive and Gram-negative bacteria
  • Potent brain penetrant neutral sphingomyelinase (N-SMase) inhibitor