Learn More
Medchemexpress LLC Omeprazole | 73590-58-6 | 99.9% | 345.42 | 100 MG
SDP
Catalog No. 5000365060
Encompass_Preferred
Omeprazole is a proton pump inhibitor (PPI) used for treating acid-related gastrointestinal disorders. It competitively inhibits CYP2C19 activity and inhibits the growth of both Gram-positive and Gram-negative bacteria. Additionally, it is a potent brain penetrant neutral sphingomyelinase (N-SMase) inhibitor and an exosome inhibitor.
  • Proton pump inhibitor (PPI)
  • Competitively inhibits CYP2C19 activity
  • Inhibits growth of Gram-positive and Gram-negative bacteria
  • Potent brain penetrant neutral sphingomyelinase (N-SMase) inhibitor
  • Exosome inhibitor
  • Inhibits gastric acid secretion
  • Acid-labile compound
Catalog No. 50-003-65060 Supplier Medchemexpress LLC Supplier No. 100MGHYB0113
Omeprazole is a proton pump inhibitor (PPI) used for treating acid-related gastrointestinal disorders. It competitively inhibits CYP2C19 activity and inhibits the growth of both Gram-positive and Gram-negative bacteria. Additionally, it is a potent brain penetrant neutral sphingomyelinase (N-SMase) inhibitor and an exosome inhibitor.
  • Proton pump inhibitor (PPI)
  • Competitively inhibits CYP2C19 activity
  • Inhibits growth of Gram-positive and Gram-negative bacteria
  • Potent brain penetrant neutral sphingomyelinase (N-SMase) inhibitor
  • Exosome inhibitor
  • Inhibits gastric acid secretion
  • Acid-labile compound