Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Omeprazole | 73590-58-6 | 99.9% | 345.42 g/mol | C17H19N3O3S | 10 MG
SDP

Catalog No. 5000297200
Encompass_Preferred

Omeprazole (Standard) is an analytical reference standard of omeprazole supplied as a purified solid for research and analytical applications. It is intended for use in assay calibration, method development, and quality control, and is documented with a certificate of analysis.

  • High reported purity (99.9%).
  • Chemical formula C17H19N3O3S and molecular weight 345.42 g/mol.
  • Provided as a 10 MG pack for analytical use.
  • Suitable for assay calibration and method development.
  • Characterized by a downloadable datasheet and COA.

Catalog No. 50-002-97200 Supplier Medchemexpress LLC Supplier No. HYB0113R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Omeprazole (Standard) is an analytical reference standard of omeprazole supplied as a purified solid for research and analytical applications. It is intended for use in assay calibration, method development, and quality control, and is documented with a certificate of analysis.

  • High reported purity (99.9%).
  • Chemical formula C17H19N3O3S and molecular weight 345.42 g/mol.
  • Provided as a 10 MG pack for analytical use.
  • Suitable for assay calibration and method development.
  • Characterized by a downloadable datasheet and COA.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.