Learn More
Medchemexpress LLC Omeprazole | 73590-58-6 | 99.96% | C17H19N3O3S | 5 G
SDP
Catalog No. 5000432430
Encompass_Preferred
Omeprazole (H 16868) is a proton pump inhibitor (PPI) primarily used for treating acid-related gastrointestinal disorders. It functions by competitively inhibiting CYP2C19 activity and suppressing gastric acid secretion. Additionally, this compound exhibits antibacterial properties against Gram-positive and Gram-negative bacteria and acts as a brain-penetrant neutral sphingomyelinase (N-SMase) inhibitor.
  • Treats dyspepsia, peptic ulcer disease, gastroesophageal reflux disease, laryngopharyngeal reflux, and Zollinger-Ellison syndrome.
  • Inhibits growth of Gram-positive and Gram-negative bacteria.
  • Functions as a proton pump inhibitor.
  • Competitively inhibits CYP2C19 activity.
  • Inhibits gastric acid secretion.
  • Acts as a potent brain penetrant neutral sphingomyelinase (N-SMase) inhibitor.
Catalog No. 50-004-32430 Supplier Medchemexpress LLC Supplier No. HYB01135G
Omeprazole (H 16868) is a proton pump inhibitor (PPI) primarily used for treating acid-related gastrointestinal disorders. It functions by competitively inhibiting CYP2C19 activity and suppressing gastric acid secretion. Additionally, this compound exhibits antibacterial properties against Gram-positive and Gram-negative bacteria and acts as a brain-penetrant neutral sphingomyelinase (N-SMase) inhibitor.
  • Treats dyspepsia, peptic ulcer disease, gastroesophageal reflux disease, laryngopharyngeal reflux, and Zollinger-Ellison syndrome.
  • Inhibits growth of Gram-positive and Gram-negative bacteria.
  • Functions as a proton pump inhibitor.
  • Competitively inhibits CYP2C19 activity.
  • Inhibits gastric acid secretion.
  • Acts as a potent brain penetrant neutral sphingomyelinase (N-SMase) inhibitor.