Learn More
Selleck Chemical LLC Omeprazole S1389-1g
SDP
Catalog No. 503108718
Encompass_Preferred
Omeprazole is a proton pump inhibitor that blocks H( )-K( )-ATPase used to treat dyspepsia peptic ulcer disease gastroesophageal reflux disease laryngopharyngeal reflux and Zollinger Ellison syndrome
Catalog No. 50-310-8718 Supplier Selleck Chemical LLC Supplier No. S13891G
Omeprazole is a proton pump inhibitor that blocks H( )-K( )-ATPase used to treat dyspepsia peptic ulcer disease gastroesophageal reflux disease laryngopharyngeal reflux and Zollinger Ellison syndrome