Learn More
Medchemexpress LLC Omeprazole-d3-1 | 934293-92-2 | 95.1% | C17H16D3N3O3S | 5 MG
SDP
Catalog No. 5000432651
Encompass_Preferred
Omeprazole-d3-1 is the deuterium labeled Omeprazole. This proton pump inhibitor (PPI) is used for the treatment of acid-related gastrointestinal disorders. It competitively inhibits CYP2C19 activity with a Ki of 2 to 6 μM, inhibits the growth of Gram-positive and Gram-negative bacteria, and acts as a potent brain penetrant neutral sphingomyelinase (N-SMase) inhibitor.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-32651 Supplier Medchemexpress LLC Supplier No. HYB0113S15MG
Omeprazole-d3-1 is the deuterium labeled Omeprazole. This proton pump inhibitor (PPI) is used for the treatment of acid-related gastrointestinal disorders. It competitively inhibits CYP2C19 activity with a Ki of 2 to 6 μM, inhibits the growth of Gram-positive and Gram-negative bacteria, and acts as a potent brain penetrant neutral sphingomyelinase (N-SMase) inhibitor.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS