Learn More
Cayman Chemical Omeprazole-d3, Purity: ≥99% deuterated forms (d1-d3); ≤1% d0, CAS: 934293-92-2, 1 mg
SDP
Catalog No. 502964278
Encompass_Preferred
An internal standard for the quantification of omeprazole by GC- or LC-MS
Catalog No. 50-296-4278 Supplier Cayman Chemical Supplier No. 220871 MG
An internal standard for the quantification of omeprazole by GC- or LC-MS