Learn More
Cayman Chemical Omeprazole-d3 500ug
SDP
Catalog No. 502987564
Encompass_Preferred
An internal standard for the quantification of omeprazole by GC- or LC-MS
Catalog No. 50-298-7564 Supplier Cayman Chemical Supplier No. 22087500 UG
An internal standard for the quantification of omeprazole by GC- or LC-MS