Learn More
Medchemexpress LLC Omeprazole-d3 | 922731-01-9 | 98.99% | C17H16D3N3O3S | 10 MM/1 ML
SDP
Catalog No. 5000432920
Encompass_Preferred
Omeprazole-d3 is a deuterium labeled form of Omeprazole, a proton pump inhibitor (PPI) used for acid-related gastrointestinal disorders. It competitively inhibits CYP2C19 activity (Ki of 2 to 6 μM) and also inhibits the growth of Gram-positive and Gram-negative bacteria. This compound can be used as a tracer and an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium labeled compound
  • Acts as a proton pump inhibitor
  • Inhibits CYP2C19 activity
  • Inhibits growth of Gram-positive and Gram-negative bacteria
  • Suitable as a tracer for quantitative analysis
  • Functions as an internal standard for quantitative analysis
Catalog No. 50-004-32920 Supplier Medchemexpress LLC Supplier No. HYB0113S10MM/1ML
Omeprazole-d3 is a deuterium labeled form of Omeprazole, a proton pump inhibitor (PPI) used for acid-related gastrointestinal disorders. It competitively inhibits CYP2C19 activity (Ki of 2 to 6 μM) and also inhibits the growth of Gram-positive and Gram-negative bacteria. This compound can be used as a tracer and an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium labeled compound
  • Acts as a proton pump inhibitor
  • Inhibits CYP2C19 activity
  • Inhibits growth of Gram-positive and Gram-negative bacteria
  • Suitable as a tracer for quantitative analysis
  • Functions as an internal standard for quantitative analysis