Learn More
Medchemexpress LLC Omeprazole-d3 | 922731-01-9 | 99.0% | 348.43 g/mol | C17H16D3N3O3S | 10 MG
SDP
Catalog No. 5000297255
Encompass_Preferred
Omeprazole-d3 is a deuterium-labeled omeprazole supplied as a stable-isotope reference standard for analytical and research applications, especially as an internal standard in LC-MS, pharmacokinetic, and metabolic studies. It is provided as a light brown solid with high chemical purity suitable for quantitative analysis.
  • Deuterium-labeled internal standard for LC-MS quantitation.
  • High chemical purity (≈99.0%) suitable for analytical applications.
  • Solid form with light brown to brown appearance for easy handling.
  • Includes CAS number 922731-01-9 for unambiguous identification.
  • Stable when stored as powder at -20°C for up to 3 years.
Catalog No. 50-002-97255 Supplier Medchemexpress LLC Supplier No. HYB0113S10MG
Omeprazole-d3 is a deuterium-labeled omeprazole supplied as a stable-isotope reference standard for analytical and research applications, especially as an internal standard in LC-MS, pharmacokinetic, and metabolic studies. It is provided as a light brown solid with high chemical purity suitable for quantitative analysis.
  • Deuterium-labeled internal standard for LC-MS quantitation.
  • High chemical purity (≈99.0%) suitable for analytical applications.
  • Solid form with light brown to brown appearance for easy handling.
  • Includes CAS number 922731-01-9 for unambiguous identification.
  • Stable when stored as powder at -20°C for up to 3 years.