Learn More
Medchemexpress LLC Omeprazole-d3 | 922731-01-9 | 99.0% | C17H16D3N3O3S | 1 MG
SDP
Catalog No. 5000432648
Encompass_Preferred
Omeprazole-d3 (H 16868-d3) is a deuterium-labeled version of Omeprazole. Omeprazole is a proton pump inhibitor (PPI) used in the treatment of acid-related gastrointestinal disorders, competitively inhibiting CYP2C19 activity and the growth of both Gram-positive and Gram-negative bacteria. The incorporation of stable heavy isotopes into drug molecules, such as deuterium, can influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Suitable for treatment of acid-related gastrointestinal disorders
  • Inhibits growth of Gram-positive and Gram-negative bacteria
Catalog No. 50-004-32648 Supplier Medchemexpress LLC Supplier No. HYB0113S1MG
Omeprazole-d3 (H 16868-d3) is a deuterium-labeled version of Omeprazole. Omeprazole is a proton pump inhibitor (PPI) used in the treatment of acid-related gastrointestinal disorders, competitively inhibiting CYP2C19 activity and the growth of both Gram-positive and Gram-negative bacteria. The incorporation of stable heavy isotopes into drug molecules, such as deuterium, can influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Suitable for treatment of acid-related gastrointestinal disorders
  • Inhibits growth of Gram-positive and Gram-negative bacteria