Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Omeprazole Impurity B Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012454 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
30 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2454 30 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2454 Supplier MilliporeSigma Supelco Supplier No. PHR198730MG
Add to Cart
Add to Cart
Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

CAS 110374-16-8
Linear Formula C16H17N3O2 S
Synonym 2-[(RS)-[(3,5-Dimethylpyridin-2-yl)methyl]sulfinyl]-5-methoxy-1 H-benzimidazole; 2-[(RS)-[(3,5-dimethylpyridin-2-yl)methyl]sulphinyl]-5-methoxy-1 h-benzimidazole; 2-[[(3,5-Dimethyl-2-pyridinyl)methyl]sulfinyl]--6-methoxy-1 H-benzimidazol; 4-Desmethoxy Omeprazole
Type Certified Reference Material
Quantity 30 mg
Formula Weight 315.39
Grade Certified Reference Material
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.