Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Omeprazole Impurity C Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015382 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
30 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5382 30 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5382 Supplier MilliporeSigma Supelco Supplier No. PHR198830MG
Add to Cart
Add to Cart
Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Omeprazole Impurity C is found to be as an impurity of omeprazole. Omeprazole is a benzimidazole derivative, which is widely used as an antiulcer drug and against other acid-related diseases.
TRUSTED_SUSTAINABILITY

Specifications

CAS 73590-85-9
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C17H19N3O2 S
MDL Number MFCD00869021
Synonym Omeprazole Sulfide; 5-METHOXY-2-[[(4-METHOXY-3,5-DIMETHYLPYRIDIN-2-YL)METHYL]SULPHANYL]-1 H-BENZIMIDAZOLE; 5-Methoxy-2-{[(4-methoxy-3,5-dimethyl-2-pyridinyl)methyl]thio}-1 H-benzimidazole; 6-Methoxy-2-{[(4-methoxy-3,5-dimethyl-2-pyridinyl)methyl]thio}-1 H-benzimidazole; Pyrmetazol; Ufiprazole
Type Certified Reference Material
Quantity 30 mg
Formula Weight 329.42
Grade Certified Reference Material
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.